missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Myeloperoxidase (MPO) Monoclonal Antibody (MPO455-8E6), eFluor™ 450, eBioscience™
GREENER_CHOICE

Catalog No. 50246003
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50246003 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50246003 Supplier Invitrogen™ Supplier No. 48129942
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

Description: The monoclonal antibody MPO455-8E6 recognizes myeloperoxidase (MPO), a protein within the azurophilic granules of myeloid cells. MPO is a multimeric protein comprised of two 55 kDa subunits and two 15 kDa subunits. The larger subunits associate with a heme protein resulting in a greenish color. As an enzyme, MPO breaks down hydrogen peroxide and oxidizes tyrosine. The products of this reaction, hypochlorous acid and tyrosyl radical, causethe cytotoxic and killing effects characteristic of neutrophils. Myeloperoxidase is important in the diagnosis of some cancers and increases in serum levels have been shown to correlate with cardiac events. Applications Reported: This MPO455-8E6 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MPO455-8E6 antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of normal human peripheral blood cells using the Intracellular Fixation & Permeablization Buffer Set (Product # 88-8824-00). For best results, whole blood should first be stained with antibodies to surface antigens then treated with 1X RBC lysis buffer (Product # 00-4333-μ) to lyse erythrocytes. Then, for intracellular staining for MPO, cells should be fixed with IC Fixation Buffer (Product # 00-8222-49) washed two times with Permeabilization Buffer (Product # 00-8333-56) and then incubated with MPO455-8E6 for 30-60 minutes. After washing, cells may be analyzed on a flow cytom...

Myeloperoxidase (MPO) is a heme protein that is synthesized during myeloid differentiation and is a major component of neutrophil azurophilic granules. It is produced as a single chain precursor and subsequently cleaved into a light and heavy chain. The mature MPO is a tetramer composed of 2 light chains and 2 heavy chains. This enzyme is responsible for producing hypohalous acids that are central to the microbicidal activity of neutrophils. Diseases that are associated with MPO include Myeloperoxidase Deficiency and Alzheimer's Disease, Familial, 1.

Specifications

Antigen Myeloperoxidase (MPO)
Applications Flow Cytometry
Classification Monoclonal
Clone MPO455-8E6
Concentration 5 μL/Test
Conjugate eFluor 450
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene MPO
Gene Accession No. P05164
Gene Alias 84 kDa myeloperoxidase; 89 kDa myeloperoxidase; mKIAA4033; MPO; Myeloperoxidase; Myeloperoxidase heavy chain; Myeloperoxidase light chain
Gene Symbols MPO
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4353
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less