missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mycoplasma pneumoniae Polyclonal Antibody

Description
PA1-7232 detects Mycoplasma pneumoniae. PA1-7232 has been successfully used in immunofluorescence procedures. The PA1-7232 immunogen is purified Mycoplasma pneumoniae.
Although they are the simplest and smallest self-replicating bacteria, Mycoplasma infections tend to be persistent, difficult to detect, diagnose, and cure. Mycoplasmas are surface parasites of human respiratory and urogenital tracts. Mycoplasma pneumoniae is often the causative agent of community-acquired pneumonia. The organism's affinity for respiratory epithelial cells and its ability to secrete hydrogen peroxide are a potent combination, leading to oxidative tissue damage.
Specifications
Specifications
| Antigen | Mycoplasma pneumoniae |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | Bacterial pneumonia; M. pneumoniae; mycoplasmen |
| Host Species | Rabbit |
| Immunogen | Purified Mycoplasma pneumoniae. |
| Purification Method | Protein A |
| Show More |