Learn More
Invitrogen™ Mycoplasma bovis Monoclonal Antibody (MY11-320.1), Invitrogen™
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA546159
 View more versions of this product
Catalog No. PIMA546159
Only null left
Description
Description
MA1-10794 specifically detects Mycoplasma bovis, and will not cross-react with Mycoplasma bovigentalium, Mycoplasma californicum, Strep. Agalactiae, Staph. Aureus, or E. coli. MA1-10794 has been successfully used in ELISA applications. The MA1-10794 immunogen is Mycoplasma bovis cells.
This antibody specifically recognize Mycoplasma bovis. It does not cross-react with Mycoplasma genitalium, Mycoplasma californicum, Strep. agalactiae, Staph. aureus or E. coli.
Specifications
Chemical Identifiers
Mycoplasma bovis
1.3 mg/mL
Monoclonal
Liquid
RUO
PBS with no preservative; pH 7.4
Mycoplasma bovis cells.
Primary
-20°C, Avoid Freeze/Thaw Cycles
MY11-320.1
ELISA
Unconjugated
Mouse
Bacteria
m bovis; M. bovis; mycoplasmen
IgG1
Protein G
Antibody
Specifications
Mycoplasma bovis
Monoclonal
1.3 mg/mL
PBS with no preservative; pH 7.4
Mouse
Protein G
RUO
Bacteria
Antibody
IgG1
ELISA
MY11-320.1
Unconjugated
m bovis; M. bovis; mycoplasmen
Mycoplasma bovis cells.
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions