missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mycoplasma bovis Monoclonal Antibody (MY11-320.1), Invitrogen™

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA110794
Description
MA1-10794 specifically detects Mycoplasma bovis, and will not cross-react with Mycoplasma bovigentalium, Mycoplasma californicum, Strep. Agalactiae, Staph. Aureus, or E. coli. MA1-10794 has been successfully used in ELISA applications. The MA1-10794 immunogen is Mycoplasma bovis cells.
This antibody specifically recognize Mycoplasma bovis. It does not cross-react with Mycoplasma genitalium, Mycoplasma californicum, Strep. agalactiae, Staph. aureus or E. coli.Chemical Identifiers
| Mycoplasma bovis | |
| 1.3 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.4 | |
| Mycoplasma bovis cells. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles |
| MY11-320.1 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Bacteria | |
| m bovis; M. bovis; mycoplasmen | |
| IgG1 | |
| Protein G | |
| Antibody |
Specifications
| Mycoplasma bovis | |
| Monoclonal | |
| 1.3 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
| ELISA | |
| MY11-320.1 | |
| Unconjugated | |
| m bovis; M. bovis; mycoplasmen | |
| Mycoplasma bovis cells. | |
| 500 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |