missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mycoplasma bovis Monoclonal Antibody (MY10-110.6), Invitrogen™
Description
MA1-10791 detects Mycoplasma bovis. MA1-10791 has been successfully used in ELISA applications. The MA1-10791 immunogen is Mycoplasma bovis cells.
This antibody specifically recognize Mycoplasma bovis. It does not cross-react with Mycoplasma genitalium, Mycoplasma californicum, Strep. agalactiae, Staph. aureus or E. coli.
Specifications
Specifications
| Antigen | Mycoplasma bovis |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | MY10-110.6 |
| Concentration | 1.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.0 |
| Gene Alias | m bovis; M. bovis; mycoplasmen |
| Host Species | Mouse |
| Immunogen | Mycoplasma bovis cells. |
| Show More |