missing translation for 'onlineSavingsMsg'
Learn More

Mycophenolate Mofetil 98.0+%, TCI America™

Catalog No. p-7133198
Change view
Click to view available options
Quantity:
1 g
100 mg
Catalog No. Quantity
M2387100MG 100 mg
M23871G 1 g
2 options

Catalog No. M2387100MG

Supplier: TCI America M2387100MG

Only null left

Chemical Identifiers

CAS 128794-94-5
Molecular Formula C23H31NO7
Molecular Weight (g/mol) 433.501
MDL Number MFCD00867568
InChI Key RTGDFNSFWBGLEC-SYZQJQIISA-N
Synonym mycophenolate mofetil, cellcept, munoloc, myfenax, mycophenolic acid morpholinoethyl ester, tm-mmf, mycophenylate mofetil, mycophenolatemofetil, cellcept tn, unii-9242ecw6r0
PubChem CID 5281078
ChEBI CHEBI:8764
IUPAC Name 2-morpholin-4-ylethyl (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoate
SMILES CC1=C(C(=C(C2=C1COC2=O)O)CC=C(C)CCC(=O)OCCN3CCOCC3)OC

Specifications

Melting Point 96°C
Color White
Quantity 100 mg
Formula Weight 433.50
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Mycophenolate Mofetil
RTECSNumber MP7746700
TSCA No