missing translation for 'onlineSavingsMsg'
Learn More

Mycophenolate mofetil, 98%

Catalog No. AC459370010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459370010 1 g
AC459370050 5 g
AC459370250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC459370010 Supplier Thermo Scientific Chemicals Supplier No. 459370010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 128794-94-5 | C23H31NO7 | 433.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 128794-94-5
Molecular Formula C23H31NO7
Molecular Weight (g/mol) 433.5
InChI Key RTGDFNSFWBGLEC-SYZQJQIISA-N
Synonym mycophenolate mofetil, cellcept, munoloc, myfenax, mycophenolic acid morpholinoethyl ester, tm-mmf, mycophenylate mofetil, mycophenolatemofetil, cellcept tn, unii-9242ecw6r0
PubChem CID 5281078
ChEBI CHEBI:8764
IUPAC Name 2-morpholin-4-ylethyl (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoate
SMILES CC1=C(C(=C(C2=C1COC2=O)O)CC=C(C)CCC(=O)OCCN3CCOCC3)OC

Specifications

Melting Point 95.0°C to 96.0°C
Ignition Residue 0.1% max
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 433.5
Percent Purity 98%
Chemical Name or Material Mycophenolate mofetil
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid release to the environment.
Collect spillage.
Dispose of contents/

RUO – Research Use Only