missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MYADM Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144100 Supplier Invitrogen™ Supplier No. PA5144100
Only null left

Rabbit Polyclonal Antibody

MYADM Polyclonal Antibody for ICC/IF, IHC (P), ELISA

MYADM, belonging to the MAL family, is a 322-amino acid protein having 7 hydrophobic transmembrane domains, several potential consensus sites for phosphorylation and sequence homology with the T-cell differentiation protein MAL and plasmolipin. It shares 75.8% sequence identity with mouse in the transmembrane regions and is associated with differentiation of hematopoietic cells or acute promyelocytic leukemia cells. Its expression is up-regulated in NB4 cells induced by all-trans retinoic acid whereas down regulation inhibits colony formation of bone marrow cells. MYADM could serve as a new marker for hematopoietic differentiation and is a target for diagnostic, prognostic and/or therapeutic interventions. It is known to express widely in brain and pancreatic islet cells.

Specifications

Antigen MYADM
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene MYADM
Gene Accession No. Q96S97
Gene Alias MYADM; myeloid associated differentiation marker; myeloid upregulated protein; myeloid up-regulated protein; myeloid-associated differentiation marker; Protein SB135; SB135; UNQ553/PRO1110
Gene Symbols MYADM
Host Species Rabbit
Immunogen Recombinant Human Myeloid-associated differentiation marker protein (1-40AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91663
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less