missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MUT Recombinant Rabbit Monoclonal Antibody (23GB4555)
GREENER_CHOICE

Catalog No. MA553766
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553766 100 μL
MA553767 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553766 Supplier Invitrogen™ Supplier No. MA553766
Only null left

Rabbit Recombinant Monoclonal Antibody

MUT Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

The product of this gene is a Golgi enzyme catalyzing an essential step in the conversion of oligomannose to complex N-glycans. The enzyme has the typical glycosyltransferase domains: a short N-terminal cytoplasmic domain, a hydrophobic non-cleavable signal-anchor domain, and a C-terminal catalytic domain. Mutations in this gene may lead to carbohydrate-deficient glycoprotein syndrome, type II. The coding region of this gene is intronless. Transcript variants with a spliced 5' UTR may exist, but their biological validity has not been determined.

Specifications

Antigen MUT
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4555
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MMUT
Gene Accession No. P16332, P22033
Gene Alias D230010K02Rik; Mcm; methylmalonyl CoA mutase; methylmalonyl Coenzyme A mutase; methylmalonyl-CoA isomerase; methylmalonyl-CoA mutase; methylmalonyl-CoA mutase, mitochondrial; methylmalonyl-Coenzyme A mutase; MMUT; MUT; mutant methylmalonyl CoA mutase; truncated methylmalonyl CoA mutase
Gene Symbols MMUT
Host Species Rabbit
Immunogen A synthesized peptide derived from human MMUT (45-95AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17850, 4594, 688517
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less