missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MURF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554058
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554058 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554058 Supplier Invitrogen™ Supplier No. PA554058
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: MEEPQMALYL QQAKELINKV GAMSKVELAG RPEPGYESME QFTVRVEHVA EMLRTIDFQP GASGEEEEVA PDGEEGSAGP EEERPDG Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 88%.

The protein encoded by this gene contains a RING finger motif and is highly similar to the ring finger proteins RNF28/MURF1 and RNF29/MURF2. In vitro studies demonstrated that this protein, RNF28, and RNF29 form heterodimers, which may be important for the regulation of titin kinase and microtubule-dependent signal pathways in striated muscles. Alternatively spliced transcript variants encoding distinct isoforms have been reported.

Specifications

Antigen MURF3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM54
Gene Accession No. Q9BYV2
Gene Alias 4930486E09Rik; 4930566I02Rik; MURF; MURF3; MuRF-3; Muscle-specific RING finger protein; Muscle-specific RING finger protein 3; muscle-specific RING-finger protein; muscle-specific RING-finger protein 3; OTTHUMP00000122598; ring finger protein 30; Rnf30; TRIM54; tripartite motif containing 54; tripartite motif-containing 54; tripartite motif-containing protein 54
Gene Symbols TRIM54
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MURF3. Recombinant protein control fragment (Product #RP-92301).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57159
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less