Learn More
Murexide
CAS: 3051-09-0 | C8H3N5O6-2 | 265.141 g/mol
Supplier: Thermo Scientific Chemicals A1754018
Description
Murexide is used as a complexometric indicator in analytical chemistry for complexometric titrations of calcium, copper nickel and cobalt. It is an inhibitor of dihydrofolate reductase. Further, it acts as an enhancer of sonochemical destruction of chlorinated hydrocarbon pollutants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMurexide is used as a complexometric indicator in analytical chemistry for complexometric titrations of calcium, copper nickel and cobalt. It is an inhibitor of dihydrofolate reductase. Further, it acts as an enhancer of sonochemical destruction of chlorinated hydrocarbon pollutants.
Solubility
Soluble in water and 1M sodium hydroxide solution. Insoluble in alcohol.
Notes
Incompatible with strong oxidizing agents. Â Â Â Â Â Â Â Â Â
Specifications
| Murexide | |
| 3051-09-0 | |
| MFCD00012777 | |
| 14,6306 | |
| Soluble in water and 1M sodium hydroxide solution. Insoluble in alcohol. | |
| C1(=C(N=C(NC1=O)[O-])[O-])N=C2C(=O)NC(=O)NC2=O | |
| 265.141 | |
| 284.19 |
| >300°C | |
| C8H3N5O6-2 | |
| 3582175 | |
| murexide, ammonium purpurate acid, 6-oxo-5-2,4,6-trioxo-1,3-diazinan-5-ylidene amino-1h-pyrimidine-2,4-diolate, 5-hexahydro-2,4,6-trioxo-5-pyrimidinyl imino-2,4,6 1h,3h,5h-pyrimidinetrione ammonium salt, 6-oxidanylidene-5-2,4,6-tris oxidanylidene-1,3-diazinan-5-ylidene amino-1h-pyrimidine-2,4-diolate | |
| MUMMBXDKKJEASE-UHFFFAOYSA-L | |
| 6-oxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1H-pyrimidine-2,4-diolate | |
| 51381221 | |
| 50 g |
Chemical Identifiers
| 3051-09-0 | |
| 265.141 | |
| MUMMBXDKKJEASE-UHFFFAOYSA-L | |
| 51381221 | |
| C1(=C(N=C(NC1=O)[O-])[O-])N=C2C(=O)NC(=O)NC2=O |
| C8H3N5O6-2 | |
| MFCD00012777 | |
| murexide, ammonium purpurate acid, 6-oxo-5-2,4,6-trioxo-1,3-diazinan-5-ylidene amino-1h-pyrimidine-2,4-diolate, 5-hexahydro-2,4,6-trioxo-5-pyrimidinyl imino-2,4,6 1h,3h,5h-pyrimidinetrione ammonium salt, 6-oxidanylidene-5-2,4,6-tris oxidanylidene-1,3-diazinan-5-ylidene amino-1h-pyrimidine-2,4-diolate | |
| 6-oxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1H-pyrimidine-2,4-diolate |
Safety and Handling
EINECSNumber : 221-266-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only