missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Murexide
Description
Murexide is used as a complexometric indicator in analytical chemistry for complexometric titrations of calcium, copper nickel and cobalt. It is an inhibitor of dihydrofolate reductase. Further, it acts as an enhancer of sonochemical destruction of chlorinated hydrocarbon pollutants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Murexide is used as a complexometric indicator in analytical chemistry for complexometric titrations of calcium, copper nickel and cobalt. It is an inhibitor of dihydrofolate reductase. Further, it acts as an enhancer of sonochemical destruction of chlorinated hydrocarbon pollutants.
Solubility
Soluble in water and 1M sodium hydroxide solution. Insoluble in alcohol.
Notes
Incompatible with strong oxidizing agents.
Murexide is used as a complexometric indicator in analytical chemistry for complexometric titrations of calcium, copper nickel and cobalt. It is an inhibitor of dihydrofolate reductase. Further, it acts as an enhancer of sonochemical destruction of chlorinated hydrocarbon pollutants.
Solubility
Soluble in water and 1M sodium hydroxide solution. Insoluble in alcohol.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 10 g |
| Beilstein | 3582175 |
| Merck Index | 14,6306 |
| Solubility Information | Soluble in water and 1M sodium hydroxide solution. Insoluble in alcohol. |
| Formula Weight | 284.19 |
| Chemical Name or Material | Murexide |
Safety and Handling
| EINECSNumber | 221-266-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only