missing translation for 'onlineSavingsMsg'
Learn More
Learn More
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18613995
|
N/A | 5 g | Amber Glass Bottle |
Each for $293.50
|
|
||||
|
18611829
|
N/A | 25 g | Amber Glass Bottle |
Each for $642.03
|
|
||||
|
18611828
|
N/A | 100 g | Amber Glass Bottle |
Each for $1,845.74
|
|
||||
|
18611827
|
N/A | 1 kg | Amber Glass Bottle |
Each for $5,628.15
|
|
||||
Chemical Identifiers
| 3051-09-0 |
Specifications
| 3051-09-0 | |
| Reagent | |
| Purple/Red to Red | |
| Murexide |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.