missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Murexide Indicator, Dry Form in NaCl, for Calcium, Certified, LabChem™
Supplier: LabChem LC175109
Specifications
| Murexide Indicator | |
| 7647-14-5,3051-09-0 | |
| ClNa | |
| MFCD00003477 | |
| Passes Test | |
| [Na+].[Cl-] | |
| 58.44 | |
| CHEBI:26710 | |
| Certified | |
| Passes Test | |
| Pink | |
| Crystalline Powder |
| Dry Form in NaCl, for Calcium | |
| 99.8,0.2 | |
| C8H8N6O6 | |
| Soluble in water | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 5234 | |
| 284.18 | |
| Poly Bottle | |
| Nitrogen oxides; Carbon monoxide; Carbon dioxide | |
| 100 g |
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| CHEBI:26710 | |
| [Na+].[Cl-] |
| ClNa | |
| MFCD00003477 | |
| 5234 | |
| sodium chloride |
Safety and Handling
GHS H Statement
Product is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature