missing translation for 'onlineSavingsMsg'
Learn More

Murexide Indicator, Dry Form in NaCl, for Calcium, Certified, LabChem™

Supplier:  LabChem LC175101

 View more versions of this product

Catalog No. LC175101


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Murexide Indicator
7647-14-5,3051-09-0
ClNa
MFCD00003477
Passes Test
[Na+].[Cl-]
58.44
CHEBI:26710
Certified
Passes Test
Pink
Solid
Dry Form in NaCl, for Calcium
99.8,0.2
C8H8N6O6
Soluble in water
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
5234
284.18
Poly Bottle
Nitrogen oxides; Carbon monoxide; Carbon dioxide
500 g

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
CHEBI:26710
[Na+].[Cl-]
ClNa
MFCD00003477
5234
sodium chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Product is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions