missing translation for 'onlineSavingsMsg'
Learn More

Murexide, ACS Reagent (Ammonium Purpurate), Reagents

Catalog No. 01337884
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337884 25 g
01337883 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337884 Supplier Labchem Inc Supplier No. C125198025C1
Only null left

Murexide, ACS Reagent (Ammonium Purpurate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 3051-09-0
Molecular Formula C8H8N6O6
Molecular Weight (g/mol) 284.19
InChI Key PCCZWUCAYILEMX-UHFFFAOYSA-O
Synonym Ammonium Purpurate, Color Index 56085
IUPAC Name ammonium 2,4,6-trioxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1,3-diazinan-5-ide
SMILES [NH4+].O=C1NC(=O)[C-](N=C2C(=O)NC(=O)NC2=O)C(=O)N1

Specifications

Melting Point >300°C
Boiling Point 1500°C
Grade ACS
Packaging Glass Bottle
Specific Gravity 1
pH Color Changes Pink (Excess Calcium) to Purple (Excess EDTA)
Color Red
Quantity 25 g
Physical Form Solid
Chemical Name or Material Murexide, ACS Reagent (Ammonium Purpurate)