missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Murexide, ACS Grade, LabChem™
Specifications
Specifications
| Melting Point | 300°C |
| Color | Maroon |
| CAS Max % | 100 |
| Quantity | 5 g |
| Packaging | Amber Glass |
| Physical Form | Powder |
| Linear Formula | C8H8N6O6 |
| Solubility Information | Soluble in water |
| Formula Weight | 284.18 |
| Grade | ACS |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Product is not hazardous. |
| Precautionary Statement | GHS P Statement If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes. If swallowed, get medical attention. |
| Recommended Storage | Room Temperature |