missing translation for 'onlineSavingsMsg'
Learn More

Murexide, 0.25% w/w in Sucrose, Reagents

Catalog No. 01334771
Change view
Click to view available options
Quantity:
100 mL
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334771 100 mL
01334770 250 g
01334769 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01334771 Supplier Labchem Inc Supplier No. CM040000100A6

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Murexide, 0.25% w/w in Sucrose, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 57-50-1 , 3051-09-0
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym Ammonium Purpurate, Color Index 56085
IUPAC Name (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Percent Purity 0.25%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.587
Color Pink
Quantity 100 mL
Physical Form Solid
Chemical Name or Material Murexide, 0.25% w/w in Sucrose