missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mupirocin 98.0+%, TCI America™
Chemical Identifiers
| CAS | 12650-69-0 |
|---|---|
| Molecular Formula | C26H44O9 |
| Molecular Weight (g/mol) | 500.629 |
| MDL Number | MFCD01711620 |
| InChI Key | MINDHVHHQZYEEK-HBBNESRFSA-N |
| Synonym | mupirocin, pseudomonic acid, bactroban, mupirocine, mupirocinum, centany, mupirocina, pseudomonic acid a, bactoderm, plasimine |
| PubChem CID | 446596 |
| ChEBI | CHEBI:7025 |
| IUPAC Name | 9-[(E)-4-[(2S,3R,4R,5S)-3,4-dihydroxy-5-[[(2S,3S)-3-[(2S,3S)-3-hydroxybutan-2-yl]oxiran-2-yl]methyl]oxan-2-yl]-3-methylbut-2-enoyl]oxynonanoic acid |
| SMILES | CC(C1C(O1)CC2COC(C(C2O)O)CC(=CC(=O)OCCCCCCCCC(=O)O)C)C(C)O |
Chemical Identifiers
| 12650-69-0 | |
| 500.629 | |
| MINDHVHHQZYEEK-HBBNESRFSA-N | |
| 446596 | |
| 9-[(E)-4-[(2S,3R,4R,5S)-3,4-dihydroxy-5-[[(2S,3S)-3-[(2S,3S)-3-hydroxybutan-2-yl]oxiran-2-yl]methyl]oxan-2-yl]-3-methylbut-2-enoyl]oxynonanoic acid |
| C26H44O9 | |
| MFCD01711620 | |
| mupirocin, pseudomonic acid, bactroban, mupirocine, mupirocinum, centany, mupirocina, pseudomonic acid a, bactoderm, plasimine | |
| CHEBI:7025 | |
| CC(C1C(O1)CC2COC(C(C2O)O)CC(=CC(=O)OCCCCCCCCC(=O)O)C)C(C)O |
Specifications
| 12650-69-0 | |
| White | |
| MFCD01711620 | |
| MINDHVHHQZYEEK-HBBNESRFSA-N | |
| 9-[(E)-4-[(2S,3R,4R,5S)-3,4-dihydroxy-5-[[(2S,3S)-3-[(2S,3S)-3-hydroxybutan-2-yl]oxiran-2-yl]methyl]oxan-2-yl]-3-methylbut-2-enoyl]oxynonanoic acid | |
| 446596 | |
| 500.63 | |
| Crystalline Powder |
| 78°C | |
| C26H44O9 | |
| mupirocin, pseudomonic acid, bactroban, mupirocine, mupirocinum, centany, mupirocina, pseudomonic acid a, bactoderm, plasimine | |
| CC(C1C(O1)CC2COC(C(C2O)O)CC(=CC(=O)OCCCCCCCCC(=O)O)C)C(C)O | |
| 500.629 | |
| CHEBI:7025 | |
| ≥98.0% (T) | |
| Mupirocin |
Safety and Handling
RTECSNumber : RA6907000
TSCA : No