missing translation for 'onlineSavingsMsg'
Learn More

Mupirocin 98.0+%, TCI America™

Catalog No. M295525MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M295525MG 25 mg
M2955100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M295525MG Supplier TCI America Supplier No. M295525MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12650-69-0
Molecular Formula C26H44O9
Molecular Weight (g/mol) 500.629
MDL Number MFCD01711620
InChI Key MINDHVHHQZYEEK-HBBNESRFSA-N
Synonym mupirocin, pseudomonic acid, bactroban, mupirocine, mupirocinum, centany, mupirocina, pseudomonic acid a, bactoderm, plasimine
PubChem CID 446596
ChEBI CHEBI:7025
IUPAC Name 9-[(E)-4-[(2S,3R,4R,5S)-3,4-dihydroxy-5-[[(2S,3S)-3-[(2S,3S)-3-hydroxybutan-2-yl]oxiran-2-yl]methyl]oxan-2-yl]-3-methylbut-2-enoyl]oxynonanoic acid
SMILES CC(C1C(O1)CC2COC(C(C2O)O)CC(=CC(=O)OCCCCCCCCC(=O)O)C)C(C)O

Specifications

Melting Point 78°C
Color White
Quantity 25 mg
Formula Weight 500.63
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Mupirocin
RTECSNumber RA6907000
TSCA No