missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connecticut Clean Room Corp. Munising LTD Cleanroom Paper
Description
- Economical alternative to Munising™ LP
- Less synthetic polymers, used to seal individual fibers into sheet
- No natural latex
- Autoclavable
- Free of inorganic fillers, minimizes inadvertent doping of products
- Accepts standard printing inks without skipping, smudging or smearing
- Heat resistant for use in laser printers and photocopy machines
Specifications
Specifications
| Color | White |
| Dimensions (L x W) | 8.5 x 11 in. |
| Autoclavable | Autoclavable |
| For Use With (Application) | Cleanrooms |
| Quantity | 2500 Sheets/Cs. |