missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Multiskan™ Ease Absorbance Microplate Reader

Catalog No. 5000850162
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5000850162 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5000850162 Supplier Thermo Scientific™ Supplier No. ME001000C
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Thermo Scientific™ Multiskan™ Ease Absorbance Microplate Reader is a modern visible-range absorbance reader (400–760 nm) designed to simplify both routine and advanced microplate-based assays. It supports 96- and 384-well plate formats and features an intuitive 8-inch full-color touchscreen with built-in shaking and incubation for temperature-sensitive protocols.

The Thermo Scientific™ Multiskan™ Ease Absorbance Microplate Reader is a modern visible-range absorbance reader (400–760 nm) designed to simplify both routine and advanced microplate-based assays. It supports 96- and 384-well plate formats and features an intuitive 8-inch full-color touchscreen with built-in shaking and incubation for temperature-sensitive protocols. With straightforward SkanIt™ Software integration to make absorbance reading easy, fast, and smart.

For researchers performing ELISA, protein quantification, cytotoxicity, enzyme kinetics, and other absorbance assays, the Multiskan™ Ease enables reliability, flexibility, and ease of use. Its easyworkflow integration—covering data transfer, analysis, and reporting—enhances laboratory productivity without the need for additional training. Backed by near 50 years of trusted performance, the Multiskan™ Ease combines robust functionality with the confidence of the Thermo Scientific brand.

Applications

Multiskan™ Ease accommodates a comprehensive array of analytical and biological assays:

  • ELISA immunoassays- endpoint and kinetic formats
  • Protein quantification- Bradford, Lowry, BCA, and peptide assays
  • Endotoxin assays- chromogenic detection for quality control
  • Cell cytotoxicity and proliferation – alamarBlue™, PrestoBlue™, and CyQUANT™ assays
  • Enzyme kinetic assays- activity, inhibition, and rate studies
  • Growth curve analysis for microbial populations- population monitoring and turbidity analysis

Features

  • Large 8-inch full-color touch screen with user-friendly multi-language interface that streamlines assay setup, data review, and instrument control
  • Efficient white LED light source offers exceptional stability, energy efficiency, and extended lifetime operation
  • Enhanced optical design and precision thermal control enables accurate absorbance measurements, consistent plate-to-plate performance, and long-term instrument stability
  • Compatibility with both 96- and 384-well plates with and without lid, integrated shaking, and incubation up to 45°C
  • Redesigned easy-to-change 8-positions filter wheel and durable hard-coated optical filters (3 pre-installed: 405 nm, 450 nm and 620 nm) for long-lasting operation and low maintenance.
  • USB-C and USB-A connectivity for simple data transfer; fully compatible integration with automation platforms; cloud-ready architecture
  • Intuitive SkanIt software for streamlined workflow management and comprehensive data analysis
  • Dependable performance and reliable daily operation with proprietary optics and self-diagnostics

The instrument includes a 1-year standard warranty, with extended warranty coverage and IQ/OQ/IPV services available separately. Verification plates (Cat. No. 24072800 and 24072805) are available for routine performance qualification, enabling verification of photometric accuracy, precision, linearity, wavelength accuracy, and stray light. Additional optical filters can be purchased separately to support different assay requirements and expand application flexibility.

Specifications

Product Type Microplate Reader
Cuvettes No
Detector Type Photodetector
Dimensions (L x W x H) 40 x 29.5 x 21.5 cm (15.7 x 11.6 x 8.5 in.)
Interface Touchscreen or PC Control
Incubator Temperature Ambient +4° to 45°C
Light Source LED
Measurement Speed Fast mode: ≤10 sec., Precision mode: ≤20 sec.
Measurement Technology Absorbance
Plate Type 96 and 384-well Plates
Reading Options Endpoint, Kinetic
Shaking Linear
Read-out Range Up to 4 Abs
Wavelength Selection Filters
Absorbance Range 400-760 nm
Software Type SkanIt Software
Filters 8-position filter wheel, 3 pre-installed: 405 nm, 450 nm and 620 nm
Voltage 100/240 V
Warranty 1-year Standard
Weight (Metric) 10 kg
Quantity 1 Ea.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.