missing translation for 'onlineSavingsMsg'
Learn More

OHAUS MultiPlate Genie

Catalog No. 01804369
Change view
Click to view available options
Voltage:
120 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
01804369 120 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01804369 Supplier OHAUS Supplier No. 83041326

Digital high-speed vortex mixing microplate shaker handles up to 4 microplates or up to 8 plates with the optional accessory tray.

  • The combined speed and 1 mm orbit provide consistant well to well and plate to plate uniformity, even in 384-well plates.
  • Featuring programable speed, time and continuous or pulse time, it is ideal for achieving accurate and repeatable results.
  • The Pulse feature allows rapid pulsing or timed on/off mixing for interval mixing.
  • Ideal for use in PCR Preparation, Digestions, Bead Mixing, ELISA, EIA, and Colorimetric Assay applications.

Specifications

Orbit 1 mm
Load Bearing Capacity (Metric) 2.3 kg
Speed Range 500 to 3000 rpm
Voltage 120 V
Product Type Shaker
Model MultiPlate Genie