Learn More
Multimedia Packages: Mitosis and Meiosis
Catalog No. p-3039672
Change view
Click to view available options
Type:
Teacher Package
Catalog No. Type
S90071 Teacher Package
1 options
Catalog No. S90071 Supplier Customer Channels Group (Ccg) Supplier No. SME21
Only null left
Includes
Teacher Package:
  • Six microscope slides in plastic box
  • Three overhead projector color transparencies
  • Six sketch- and worksheets
  • Brochure with explanatory text
  • Cardboard storage box
Student Package:
  • Six microscope slides in plastic box
  • Brochure with explanatory text
  • Cardboard storage box
Six selected microscope slides with accompanying brochure.
Teacher and Student Packages, set of six slides:
  • Allium, root tips, l.s. showing lateral view of all stages of mitosis, iron-hematoxyline
  • Whitefish mitosis, l.s. of embryo showing animal mitosis
  • Testis of mouse, t.s. showing spermatogenesis in all stages
  • Giant chromosomes from salivary gland of Chironomus, squash preparation special stained for chromomeres
  • Lilium, microspore mother cells, prophase of first division showing meiosis
  • Lilium, microspore mother cells, meta- or anaphase of first division, showing mitosis
Specifications
Type Teacher Package
Quantity 6