Learn More
Agilent BioTek MultiFlo FX Wash Module
GREENER_CHOICE
Catalog No. bt1260023
Change view
Click to view available options
For Use With (Equipment):
MFXP2, MFXP2R
Catalog No. For Use With (Equipment)
BT1260023 MFXP2, MFXP2R
1 options
Catalog No. BT1260023 Supplier Agilent BioTek Supplier No. 1260023
Only null left
Wash module for 6- to 384-well plates combines dispensing and washing automated on one platform
Specifications
Description External wash module
For Use With (Application) Primary/Secondary Screening Assays,Compound Storage,Genomics and Proteomics Research,ELISA,Cell Based Washing, Fixing, Staining, Volume or Concentration Normalization
Includes Wash module, pump, bottles, tubing, 96/384 wash manifold
Accuracy 96-/384-well plates, 300 μL/well: <3% CV,6-well plates, 5560 μL/well: <5% CV
Certifications/Compliance CE and TUV marked. RoHS compliant.
For Use With (Equipment) MFXP2, MFXP2R