Learn More
Agilent BioTek MultiFlo FX Microplate Dispenser Accessory, Manifold
Accurate and precise dispensing
Supplier: Agilent BioTek 7180560S
Description
- Syringe Dispenser Manifold
- 96-/384-well Plates - 2x8-tube
- Autoclavable
Specifications
| 2 x 8 Manifold | |
| CE and TUV marked. RoHS compliant. | |
| Autoclavable |
| Steel | |
| Half-area and PCR 96-well formats,Steel tips | |
| Multiflo FX, 96- and 384-well Dispensing |