missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Multi-Use Nestable Sharps Collectors
Nestable sharps containers save valuable storage space
$467.84 - $467.84
Specifications
| Closure Color | Clear |
|---|---|
| Color | Red |
| Autoclavable | Autoclavable |
| For Use With (Application) | Suitable for incineration or autoclaving, and general-purpose collectors for safe transport and final disposal |
| Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Closure Type | Quantity | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Closure Type | Quantity | Price | Quantity | ||
|
22253135
|
BD
305343 |
8 qt. | 7.6 L | Open Top | 24/Pk. |
Case of 24 Each for $467.84
|
|
||
Description
- “Point-first” drop minimizes the need to manipulate syringe when dropping it into collector
- Opening provides one-way safety valve to reduce risks of tampering, spilling, or needle pop-back
- Rigid polypropylene, general-purpose collectors with are durable and puncture-resistant for safe transport and final disposal
- Suitable for incineration or autoclaving
- Brackets available separately; see cat. no. 19065796
Specifications
| Clear | |
| Red | |
| Autoclavable | |
| Suitable for incineration or autoclaving, and general-purpose collectors for safe transport and final disposal |