missing translation for 'onlineSavingsMsg'
Learn More

BD Multi-Use Nestable Sharps Collectors

Catalog No. 22253135
Change view
Click to view available options
Capacity (English):
8 qt.
Capacity (Metric):
7.6 L
Closure Type:
Open Top
Quantity:
24/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Capacity (Metric) Closure Type Quantity
22253135 8 qt. 7.6 L Open Top 24/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22253135 Supplier BD Supplier No. 305343

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nestable sharps containers save valuable storage space

  • “Point-first” drop minimizes the need to manipulate syringe when dropping it into collector
  • Opening provides one-way safety valve to reduce risks of tampering, spilling, or needle pop-back
  • Rigid polypropylene, general-purpose collectors with are durable and puncture-resistant for safe transport and final disposal
  • Suitable for incineration or autoclaving
  • Brackets available separately; see cat. no. 19065796

Specifications

Capacity (Metric) 7.6 L
Closure Type Open Top
Closure Color Clear
Capacity (English) 8 qt.
Color Red
Quantity 24/Pk.
Autoclavable Autoclavable
For Use With (Application) Suitable for incineration or autoclaving, and general-purpose collectors for safe transport and final disposal
Material Polypropylene