missing translation for 'onlineSavingsMsg'
Learn More

Multi Ion anion IC standard solution in H2O, Specpure™

Catalog No. AA41693AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41693AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA41693AE Supplier Thermo Scientific Chemicals Supplier No. 041693AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

Multi Ion anion IC standard solution in H2O is utilized as a standard solution in analytical chemistry and ion chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Multi Ion anion IC standard solution in H2O
Name Note Br-, Cl-, F-, NO3-, PO4-3 and SO4-2at 100μg/mL. Keep refrigerated
Physical Form Liquid
MDL Number MFCD00144814
Solubility Miscible with water
Quantity 100 mL

RUO – Research Use Only