missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Multi-Element QC-7 Check Standard Solution, Specpure™, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Multi-Element QC-7 Check Standard Solution |
| Name Note | Standard Solution |
| Physical Form | Liquid |
| Molecular Formula | 5% HNO3/tr· F? |
| Concentration | Matrix: 5% HNO3/tr. F- |
| MDL Number | MFCD00210654 |
| UN Number | UN3264 |
| Type | Analytical Standard |
| Quantity | 100 mL |
| Grade | Specpure |
RUO – Research Use Only