missing translation for 'onlineSavingsMsg'
Learn More

Multi-Element QC-19 Check Standard Solution, Specpure™

$660.22 - $660.22

Specifications

Chemical Name or Material Multi-Element QC-19 Check Standard Solution
Physical Form Liquid
Concentration Matrix: 5% HNO3/tr. F-/tr. tartaric acid
MDL Number MFCD00210655
UN Number UN3264
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA39146AE
View Documents
Thermo Scientific Chemicals
039146AE
100 mL
Each for $660.22
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Multi-Element QC-19 Check Standard Solution
Matrix: 5% HNO3/tr. F-/tr. tartaric acid
UN3264
Liquid
MFCD00210655
Specpure
Videos
SDS
Documents

Documents

RUO – Research Use Only