missing translation for 'onlineSavingsMsg'
Learn More

Multi-Element QC-19 Check Standard Solution, Specpure™

Catalog No. AA39146AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39146AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA39146AE Supplier Thermo Scientific Chemicals Supplier No. 039146AE

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Multi-Element QC-19 Check Standard Solution
Physical Form Liquid
Concentration Matrix: 5% HNO3/tr. F-/tr. tartaric acid
MDL Number MFCD00210655
UN Number UN3264
Quantity 100 mL
Grade Specpure

RUO – Research Use Only