accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
EN
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
Sign In
Don't have an account?
Create an Account
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Buffers and Standards
Matrix Blanks and Standardization Solutions
AA47370AE
Print
Thermo Scientific™ Multi-Element ICH/USP Oral Standard Solution B, Specpure™
Catalog No.
AA47370AE
Change view
Chip view icon
List view icon
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
Quantity
AA47370AE
100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No.
AA47370AE
Supplier
Thermo Scientific™
Supplier No.
47370AE
Sign In
or
Register
to check your price and availability.
$335.56
/
Each
Add to Cart
Add to Cart
Description
Specifications
Product Suggestions
Videos
Documents
Certificates
Safety and Handling
Safety Services
Promotions
Description
Form:Liquid
Specifications
Specifications
Quantity
100 mL
Product Suggestions
Documents