accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
EN
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
My Account
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Buffers and Standards
Matrix Blanks and Standardization Solutions
AA473846C
Print
Thermo Scientific™ Multi-Element ICH Q3D & USP 232 Kit for Metals Analysis, Specpure™
Catalog No.
AA473846C
Change view
Chip view icon
List view icon
Click to view available options
Quantity:
5 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
Quantity
AA473846C
5 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No.
AA473846C
Supplier
Thermo Scientific™
Supplier No.
473846C
Sign In
or
Register
to check your price and availability.
Quantity
Add to Cart
Add to Cart
Description
Specifications
Product Suggestions
Videos
Documents
Certificates
Safety and Handling
Safety Services
Promotions
Description
Form:Liquid
Specifications
Specifications
Quantity
5 x 100 mL
Product Suggestions
Documents