missing translation for 'onlineSavingsMsg'
Learn More

Mubritinib 98.0+%, TCI America™

Catalog No. p-7133556
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M305825MG 25 mg
M3058100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M305825MG Supplier TCI America Supplier No. M305825MG
Only null left

Chemical Identifiers

CAS 366017-09-6
Molecular Formula C25H23F3N4O2
Molecular Weight (g/mol) 468.48
MDL Number MFCD09954135
InChI Key ZTFBIUXIQYRUNT-MDWZMJQESA-N
Synonym (E)-4-[[4-[4-(1H-1,2,3-Triazol-1-yl)butyl]phenoxy]methyl]-2-[4-(trifluoromethyl)styryl]oxazole, TAK 165
PubChem CID 6444692
IUPAC Name 4-[[4-[4-(triazol-1-yl)butyl]phenoxy]methyl]-2-[(E)-2-[4-(trifluoromethyl)phenyl]ethenyl]-1,3-oxazole
SMILES C1=CC(=CC=C1CCCCN2C=CN=N2)OCC3=COC(=N3)C=CC4=CC=C(C=C4)C(F)(F)F

Specifications

Melting Point 160°C
Quantity 25 mg
Formula Weight 468.48
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Mubritinib
TSCA No
Recommended Storage Refrigerator