missing translation for 'onlineSavingsMsg'
Learn More

Mubritinib 98.0+%, TCI America™

Catalog No. m3058100mg
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
25 mg
Catalog No. Quantity
M3058100MG 100 mg
M305825MG 25 mg
2 options

Catalog No. M3058100MG

Supplier: TCI America M3058100MG

Only null left

Chemical Identifiers

CAS 366017-09-6
Molecular Formula C25H23F3N4O2
Molecular Weight (g/mol) 468.48
MDL Number MFCD09954135
InChI Key ZTFBIUXIQYRUNT-MDWZMJQESA-N
Synonym (E)-4-[[4-[4-(1H-1,2,3-Triazol-1-yl)butyl]phenoxy]methyl]-2-[4-(trifluoromethyl)styryl]oxazole, TAK 165
PubChem CID 6444692
IUPAC Name 4-[[4-[4-(triazol-1-yl)butyl]phenoxy]methyl]-2-[(E)-2-[4-(trifluoromethyl)phenyl]ethenyl]-1,3-oxazole
SMILES C1=CC(=CC=C1CCCCN2C=CN=N2)OCC3=COC(=N3)C=CC4=CC=C(C=C4)C(F)(F)F

Specifications

Melting Point 160°C
Quantity 100 mg
Formula Weight 468.48
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Mubritinib
TSCA No
Recommended Storage Refrigerator