missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ MTT, Calbiochem™,
Membrane-permeable yellow dye that is reduced by mitochondrial reductases in living cells to form the dark blue product, MTT-formazan
Supplier: MilliporeSigma™ 47598910GM
Specifications
| MTT | |
| 195°C | |
| C18H16BrN5S | |
| 10 g | |
| AZKSAVLVSZKNRD-UHFFFAOYSA-M | |
| 2-(4,5-dimethyl-1,3-thiazol-2-yl)-3,5-diphenyl-3H-1,2λâµ,3,4-tetrazol-2-ylium bromide | |
| 64965 | |
| 414.3 | |
| Solid |
| 298-93-1 | |
| White | |
| MFCD00011964,MFCD00066662 | |
| thiazolyl blue, thiazolyl blue tetrazolium bromide, mmt tetrazolium, methylthiazoletetrazolium, mtt, thiazolyl blue monotetrazolium, thiazole blue, 3-4,5-dimethylthiazol-2-yl-2,5-diphenyltetrazolium bromide, mtt van, unii-euy85h477i | |
| [Br-].CC1=C(C)N=C(S1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1 | |
| 414.33 | |
| CHEBI:53233 | |
| ≥98% |
Chemical Identifiers
| 298-93-1 | |
| 414.33 | |
| AZKSAVLVSZKNRD-UHFFFAOYSA-M | |
| 64965 | |
| 2-(4,5-dimethyl-1,3-thiazol-2-yl)-3,5-diphenyl-3H-1,2λâµ,3,4-tetrazol-2-ylium bromide |
| C18H16BrN5S | |
| MFCD00011964,MFCD00066662 | |
| thiazolyl blue, thiazolyl blue tetrazolium bromide, mmt tetrazolium, methylthiazoletetrazolium, mtt, thiazolyl blue monotetrazolium, thiazole blue, 3-4,5-dimethylthiazol-2-yl-2,5-diphenyltetrazolium bromide, mtt van, unii-euy85h477i | |
| CHEBI:53233 | |
| [Br-].CC1=C(C)N=C(S1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1 |
Safety and Handling
EINECSNumber : 206-069-5
RTECSNumber : XF8060000
Recommended Storage : 15° to 30°C (59° to 86°F); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).