missing translation for 'onlineSavingsMsg'
Learn More

MTT, 95%, Spectrum™ Chemical

Catalog No. p-7165933
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613994 5 g Amber Glass Bottle
18613993 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18613994 Supplier Spectrum Chemical Mfg Cor Supplier No. MT1105GM
Only null left

Chemical Identifiers

CAS 298-93-1

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 0.95
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.