missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liofilchem™ MTS™ Tetracycline [TE] 0.016-256 μg/mL
MIC Test Strips are a quantitative assay for determining the Minimum Inhibitory Concentration (MIC) of antimicrobial agents against microorganisms to indicate appropriate patient treatment and for identifying resistance patterns.
$228.25 - $599.21
Specifications
| Antimicrobial Agents | Tetracycline |
|---|---|
| Certifications/Compliance | FDA Cleared |
| Concentration | 0.016 to 256 μg/mL |
| Content And Storage | -20°C or 2°C to 8°C until the given expiry date |
| Description | Liofilchem™ MTS™ Tetracycline |
| Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity & Availability | ||||
22777902
|
Liofilchem™
921141 |
10/Pk. | Individually Packed |
Pack of 10 Each for $228.25
|
|
||||
22777900
|
Liofilchem™
92114 |
30/Pk. | Individually Packed |
Pack of 30 Each for $599.21
|
|
||||
Description
Specifications
| Tetracycline | |
| 0.016 to 256 μg/mL | |
| Liofilchem™ MTS™ Tetracycline | |
| MIC Test Strip |
| FDA Cleared | |
| -20°C or 2°C to 8°C until the given expiry date | |
| TE | |
| Antimicrobial Susceptibility Testing, For determining the MIC of antimicrobial agents against microorganisms |
Non-hazardous according to current legislation, but falls within the specific field of application where a safety data sheet must be supplied because it can cause phenomena of sensitization in sensitive subjects if it comes into contact with the skin.