missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liofilchem™ MTS™ Spectinomycin [SPC] 0.064-1024 μg/mL
MIC Test Strips are a quantitative assay for determining the Minimum Inhibitory Concentration (MIC) of antimicrobial agents against microorganisms to indicate appropriate patient treatment and for identifying resistance patterns.
$269.86 - $2,184.43
Specifications
| Antimicrobial Agents | Spectinomycin |
|---|---|
| Concentration | 0.064 to 1024 μg/mL |
| Content And Storage | -20°C or 2°C to 8°C until the given expiry date |
| Description | Liofilchem™ MTS™ Spectinomycin |
| Antibiotic Code | SPC |
| Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity & Availability | ||||
22777692
|
Liofilchem™
920141 |
10/Pk. | Individually Packed |
Pack of 10 Each for $269.86
|
|
||||
22777690
|
Liofilchem™
92014 |
30/Pk. | Individually Packed |
Pack of 30 Each for $700.29
|
|
||||
22777691
|
Liofilchem™
920140 |
100/Pk. | Canister Pack |
Pack of 100 Each for $2,184.43
|
|
||||
Description
Specifications
| Spectinomycin | |
| -20°C or 2°C to 8°C until the given expiry date | |
| SPC | |
| Antimicrobial Susceptibility Testing, For determining the MIC of antimicrobial agents against microorganisms |
| 0.064 to 1024 μg/mL | |
| Liofilchem™ MTS™ Spectinomycin | |
| MIC Test Strip |
Non-hazardous according to current legislation, but falls within the specific field of application where a safety data sheet must be supplied because it can cause phenomena of sensitization in sensitive subjects if it comes into contact with the skin.