missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liofilchem™ MTS™ Rifampicin [RD] 0.016-256 μg/mL
Description
Specifications
Specifications
| Antimicrobial Agents | Rifampicin |
| Concentration | 0.016 to 256 μg/mL |
| Content And Storage | -20°C or 2°C to 8°C until the given expiry date |
| Description | Liofilchem™ MTS™ Rifampicin |
| Antibiotic Code | RD |
| Product Type | MIC Test Strip |
| For Use With (Application) | Antimicrobial Susceptibility Testing, For determining the MIC of antimicrobial agents against microorganisms |
| Packaging Type | Individually Packed |
| Quantity | 10/Pk. |
Non-hazardous according to current legislation, but falls within the specific field of application where a safety data sheet must be supplied because it can cause phenomena of sensitization in sensitive subjects if it comes into contact with the skin.