missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liofilchem™ MTS™ Plazomicin [PLZ] 0.016-256 μg/mL
MIC Test Strips are a quantitative assay for determining the Minimum Inhibitory Concentration (MIC) of antimicrobial agents against microorganisms to indicate appropriate patient treatment and for identifying resistance patterns.
$228.25 - $1,883.29
Specifications
| Antimicrobial Agents | Plazomicin |
|---|---|
| Certifications/Compliance | FDA Cleared |
| Concentration | 0.016 to 256 μg/mL |
| Content And Storage | -20°C or 2°C to 8°C until the given expiry date |
| Description | Liofilchem™ MTS™ Plazomicin |
| Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity & Availability | ||||
22777836
|
Liofilchem™
920701 |
10/Pk. | Individually Packed |
Pack of 10 Each for $228.25
|
|
||||
22777834
|
Liofilchem™
92070 |
30/Pk. | Individually Packed |
Pack of 30 Each for $599.21
|
|
||||
22777835
|
Liofilchem™
920700 |
100/Pk. | Canister Pack |
Pack of 100 Each for $1,883.29
|
|
||||
Description
Specifications
| Plazomicin | |
| 0.016 to 256 μg/mL | |
| Liofilchem™ MTS™ Plazomicin | |
| MIC Test Strip |
| FDA Cleared | |
| -20°C or 2°C to 8°C until the given expiry date | |
| PLZ | |
| Antimicrobial Susceptibility Testing, For determining the MIC of antimicrobial agents against microorganisms |
Non-hazardous according to current legislation, but falls within the specific field of application where a safety data sheet must be supplied because it can cause phenomena of sensitization in sensitive subjects if it comes into contact with the skin.