missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liofilchem™ MTS™ Kanamycin [K] 0.016-256 μg/mL
MIC Test Strips are a quantitative assay for determining the Minimum Inhibitory Concentration (MIC) of antimicrobial agents against microorganisms to indicate appropriate patient treatment and for identifying resistance patterns.
$582.89
Specifications
| Antimicrobial Agents | Kanamycin |
|---|---|
| Concentration | 0.016 to 256 μg/mL |
| Content And Storage | -20°C or 2°C to 8°C until the given expiry date |
| Description | Liofilchem™ MTS™ Kanamycin |
| Antibiotic Code | K |
Description
Specifications
| Kanamycin | |
| -20°C or 2°C to 8°C until the given expiry date | |
| K | |
| Antimicrobial Susceptibility Testing, For determining the MIC of antimicrobial agents against microorganisms |
| 0.016 to 256 μg/mL | |
| Liofilchem™ MTS™ Kanamycin | |
| MIC Test Strip |
Non-hazardous according to current legislation, but falls within the specific field of application where a safety data sheet must be supplied because it can cause phenomena of sensitization in sensitive subjects if it comes into contact with the skin.