missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liofilchem™ MTS™ Dalbavancin [DAL] 0.002-32 μg/mL
MIC Test Strips are a quantitative assay for determining the Minimum Inhibitory Concentration (MIC) of antimicrobial agents against microorganisms to indicate appropriate patient treatment and for identifying resistance patterns.
$269.86
Specifications
| Antimicrobial Agents | Dalbavancin |
|---|---|
| Certifications/Compliance | FDA Cleared |
| Concentration | 0.002 to 32 μg/mL |
| Content And Storage | -20°C or 2°C to 8°C until the given expiry date |
| Description | Liofilchem™ MTS™ Dalbavancin |
Description
Specifications
| Dalbavancin | |
| 0.002 to 32 μg/mL | |
| Liofilchem™ MTS™ Dalbavancin | |
| MIC Test Strip |
| FDA Cleared | |
| -20°C or 2°C to 8°C until the given expiry date | |
| DAL | |
| Antimicrobial Susceptibility Testing, For determining the MIC of antimicrobial agents against microorganisms |
Non-hazardous according to current legislation, but falls within the specific field of application where a safety data sheet must be supplied because it can cause phenomena of sensitization in sensitive subjects if it comes into contact with the skin.