missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liofilchem™ MTS™ Cloxacillin [CX] 0.016-256 μg/mL
MIC Test Strips are a quantitative assay for determining the Minimum Inhibitory Concentration (MIC) of antimicrobial agents against microorganisms to indicate appropriate patient treatment and for identifying resistance patterns.
Specifications
| Antimicrobial Agents | Cloxacillin |
|---|---|
| Concentration | 0.016 to 256 μg/mL |
| Content And Storage | -20°C or 2°C to 8°C until the given expiry date |
| Description | Liofilchem™ MTS™ Cloxacillin |
| Antibiotic Code | CX |
| Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity & Availability | ||||
22777780
|
Liofilchem™
92044 |
30/Pk. | Individually Packed |
N/A
|
N/A | ||||
Description
Specifications
| Cloxacillin | |
| -20°C or 2°C to 8°C until the given expiry date | |
| CX | |
| Antimicrobial Susceptibility Testing, For determining the MIC of antimicrobial agents against microorganisms |
| 0.016 to 256 μg/mL | |
| Liofilchem™ MTS™ Cloxacillin | |
| MIC Test Strip |
Non-hazardous according to current legislation, but falls within the specific field of application where a safety data sheet must be supplied because it can cause phenomena of sensitization in sensitive subjects if it comes into contact with the skin.