missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTNR1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150133
Description
MTNR1A Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes one of two high affinity forms of a receptor for melatonin, the primary hormone secreted by the pineal gland. This receptor is a G-protein coupled, 7-transmembrane receptor that is responsible for melatonin effects on mammalian circadian rhythm and reproductive alterations affected by day length. The receptor is an integral membrane protein that is readily detectable and localized to two specific regions of the brain. The hypothalamic suprachiasmatic nucleus appears to be involved in circadian rhythm while the hypophysial pars tuberalis may be responsible for the reproductive effects of melatonin.Chemical Identifiers
| MTNR1A | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O02781, P48039 | |
| 100627802, 4543 | |
| Recombinant Melatonin Receptor 1A (MTNR1A) (Gly296-Val350). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Pig | |
| MTNR1A | |
| mel1a melatonin receptor; mel1a receptor; Mel-1A-R; melatonin receptor 1A; melatonin receptor type 1A; MelR; MR; MT1; Mtnr1a | |
| MTNR1A | |
| Primary | |
| Protein A |
Specifications
| MTNR1A | |
| Polyclonal | |
| Unconjugated | |
| MTNR1A | |
| mel1a melatonin receptor; mel1a receptor; Mel-1A-R; melatonin receptor 1A; melatonin receptor type 1A; MelR; MR; MT1; Mtnr1a | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100627802, 4543 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O02781, P48039 | |
| MTNR1A | |
| Recombinant Melatonin Receptor 1A (MTNR1A) (Gly296-Val350). | |
| 200 μL | |
| Primary | |
| Human, Pig | |
| Antibody |