missing translation for 'onlineSavingsMsg'
Learn More

MTA1 Monoclonal Antibody (1F3F12), Invitrogen™

Catalog No. 390700
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
390700 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 390700 Supplier Thermo Fisher Scientific Supplier No. 390700
Only null left

Mouse Monoclonal Antibody

This antibody is specific for the C-terminal region of the human MTA1 protein. On Western blots, it identifies doublets at ∽80 kDa. A band of unknown origin is observed at ∽150 kDa.

This gene encodes a protein that was identified in a screen for genes expressed in metastatic cells, specifically, mammary adenocarcinoma cell lines. Expression of this gene has been correlated with the metastatic potential of at least two types of carcinomas although it is also expressed in many normal tissues. The role it plays in metastasis is unclear. It was initially thought to be the 70kD component of a nucleosome remodeling deacetylase complex, NuRD, but it is more likely that this component is a different but very similar protein. These two proteins are so closely related, though, that they share the same types of domains. These domains include two DNA binding domains, a dimerization domain, and a domain commonly found in proteins that methylate DNA. The profile and activity of this gene product suggest that it is involved in regulating transcription and that this may be accomplished by chromatin remodeling.

Specifications

Antigen MTA1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1F3F12
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation tissue culture supernatant with 0.1% sodium azide
Gene MTA1
Gene Accession No. Q13330, Q62599, Q8K4B0
Gene Alias metastasis associated 1; metastasis associated gene 1 protein; metastasis-associated protein MTA1; MTA1; Zg29
Gene Symbols MTA1
Host Species Mouse
Immunogen Synthetic peptide derived from the N-terminal region of the human MST3 protein
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116870, 64520, 9112
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less