missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MT1G Polyclonal Antibody
GREENER_CHOICE

Catalog No. fepa5144126
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144126 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144126 Supplier Invitrogen™ Supplier No. PA5144126
Only null left

Rabbit Polyclonal Antibody

MT1G Polyclonal Antibody for IHC (P), ELISA

Metallothioneins have a high content of cysteine residues that bind various heavy metals; these proteins are transcriptionally regulated by both heavy metals and glucocorticoids.

Specifications

Antigen MT1G
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene MT1G
Gene Accession No. P13640
Gene Alias MT1K; MT1M
Gene Symbols MT1G
Host Species Rabbit
Immunogen Recombinant Human Metallothionein-1G protein (1-59AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4495
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less