missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MT1E Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149811
Description
MT1E Polyclonal Antibody for Western Blot, IHC
MT1E is a protein coding gene. Metallothioneins have a high content of cysteine residues that bind various heavy metals; these proteins are transcriptionally regulated by both heavy metals and glucocorticoids.Specifications
| MT1E | |
| Polyclonal | |
| Unconjugated | |
| MT1E | |
| metallothionein 1E; metallothionein 1E (functional); metallothionein D; metallothionein-1E; Metallothionein-IE; MT1; MT1E; MT-1E; MTD; MT-IE | |
| Rabbit | |
| Protein A | |
| RUO | |
| 4493 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P04732 | |
| MT1E | |
| Recombinant Metallothionein 1E (MT1E) (Met1-Ala61). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody |