missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MT-ND5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536600
Description
This antibody detects endogenous protein at a molecular weight of 67 kDa. Purity is >95% by SDS-PAGE.
A small number of complex I subunits are the products of mitochondrial genes (subunits 1-7), while the remainder are nuclear encoded and imported from the cytoplasm. NADH dehydrogenase subunit 5 (ND5) is most likely a component of the hydrophobic protein fragment of Complex I. Mutations in the gene encoding for ND5 are implicated in Mitochondrial neurogastrointestinal encephalomyopathy, an autosomal recessive multisystem disorder that results in dramatically elevated levels of circulating thymidine and deoxyuridine.Chemical Identifiers
| MT-ND5 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| MT-ND5 | |
| mitochondrially encoded NADH dehydrogenase 5; MTND5; mt-Nd5; NADH Dehydrogenase 5; NADH dehydrogenase subunit 5; NADH5; NADH-ubiquinone oxidoreductase chain 5; ND5 | |
| MT-ND5, ND5 | |
| Primary | |
| Antigen affinity chromatography |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% sodium azide; pH 7.2 | |
| P03915, P03921, P11661 | |
| 17721, 26202, 4540 | |
| Synthetic peptide corresponding to amino acids 321-370 of human ND5. | |
| Antibody |
Specifications
| MT-ND5 | |
| Polyclonal | |
| Unconjugated | |
| MT-ND5 | |
| mitochondrially encoded NADH dehydrogenase 5; MTND5; mt-Nd5; NADH Dehydrogenase 5; NADH dehydrogenase subunit 5; NADH5; NADH-ubiquinone oxidoreductase chain 5; ND5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17721, 26202, 4540 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide; pH 7.2 | |
| P03915, P03921, P11661 | |
| MT-ND5, ND5 | |
| Synthetic peptide corresponding to amino acids 321-370 of human ND5. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |